
Brilliant green
TCIAB0789-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Green to Dark green powder to crystal
- Lambda max.:626.0 to 632.0 nm (EtOH (50%))
- Absorbance (E1%1cm):min. 2,000 (EtOH (50%), 626.0 to 632.0 nm) (calcd.on dried substance)
- Drying loss:max. 5.0 %
:
:482.65 g/mol
:14,01374
:211-190-1
:GHS07
:Ambient
:633-03-4
:C₂₇H₃₄N₂O₄S
:MFCD00011880