Norway
Order Entry
Cold traps, Type KF
Cold traps, Type KF
Supplier:  KGW
Certificates
undefined
Cold traps, Type KF
Supplier:  KGW
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Cold trap, KF 29-K, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    Cold trap, KF 29-K, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    478-4422
    1731
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 14 550,00
    kr 14 550,00
    1 * 1 Items
       kr 14 550,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, KF 29-OK, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    Cold trap, KF 29-OK, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    KGWA1735
    1735
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 15 275,00
    kr 15 275,00
    1 * 1 Items
       kr 15 275,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity
    Cold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity
    KGWA17371
    17371
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 16 825,00
    kr 16 825,00
    1 * 1 Items
       kr 16 825,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
    Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
    KGWA17372
    17372
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 15 875,00
    kr 15 875,00
    1 * 1 Items
       kr 15 875,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, S 29-O-A, borosilicate glass 3,3, with S 29 spherical joints and O-ring seal, 150 ml condensate capacity
    Cold trap, S 29-O-A, borosilicate glass 3,3, with S 29 spherical joints and O-ring seal, 150 ml condensate capacity
    KGWA17377
    17377
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 8 325,00
    kr 8 325,00
    1 * 1 Items
       kr 8 325,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    KGWA1740
    1740
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 14 300,00
    kr 14 300,00
    1 * 1 Items
       kr 14 300,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
    Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
    KGWA17402
    17402
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 18 425,00
    kr 18 425,00
    1 * 1 Items
       kr 18 425,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
    KGWA1741
    1741
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 6 750,00
    kr 6 750,00
    1 * 1 Items
       kr 6 750,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable