Eye shower, table-mounted
Leverandør: Broen Lab
118-1932EA 0 NOK
BROE17.200.009
Eye shower, table-mounted
Eye shower for table-mounting. With two heads in brass with chemical resistant polycoating.
  • With push handle
  • Water is regulated with a built-in FLOWFIX (26 l/min)
Min. working pressure: 1,5 bar
Weight: Approx. 2 kg
Connection: G1/2" male