Emergency showers, for wall mounting
Leverandør: Broen Lab
118-0143EA 5925 NOK
118-0143
Emergency showers, for wall mounting
Førstehjelpsutstyr Dusjer
Body shower for exposed pipework with self-draining head and pull rod. In brass with chemical resistant polycoating.
  • Quick mounting system: Easy to mount and install
  • Shower head with improved spray pattern
  • Self-draining, no dripping
  • With pull rod
  • Pre-mounted valve for body shower
Weight: ~3 kg
Connection: G ¾“
Min. working pressure: 1 bar
Sertifiseringer: EN 15154-1; ANSI Z358.1. TÜV certified