Screw caps for tubes
: DELTALAB
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
- Both models of caps are fully autoclavable and are pharmaceutical quality
- Excellent chemical resistance
- Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
- MaterialThreadPk
- 617000Aluminium15-4155001 * 500 Itemskr 4 650,00
Specifications
Environmentally Preferable - 617100PP13-4151.0001 * 1.000 Itemskr 4 100,00
Specifications
Environmentally Preferable - 617200PP15-4151.0001 * 1.000 Itemskr 4 225,00
Specifications
Environmentally Preferable - 617300PP18-4155001 * 500 Itemskr 3 300,00
Specifications
Environmentally Preferable