Cell culture media, RPMI 1640
Supplier: Biowest
Certificates
About this item
RPMI are general purpose enriched media with extensive applications for a broad spectrum of mammalian and hybridoma cells, including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes. It was originally formulated for suspension cultures and monolayer culture of human leukaemia cells.
Product Information
- Katalog #Leverandør #Media FormatPkAvailabilityPrice
- RPMI 1640 with L-glutamineRPMI 1640 with L-glutamineL0500-100L0500-100Liquid100 mL1 * 100 mLkr 275,00
- RPMI 1640 with L-glutamineRPMI 1640 with L-glutamineL0500-500Liquid500 mL1 * 500 mLkr 120,00
- RPMI 1640 without L-glutamineRPMI 1640 without L-glutamineL0501-100Liquid100 ml1 * 100 mLkr 87,00
- RPMI 1640 without L-glutamineRPMI 1640 without L-glutamineL0501-500Liquid500 ml1 * 500 mLkr 110,00
- RPMI 1640 without L-glutamine
- RPMI 1640 with stable glutamineRPMI 1640 with stable glutamineL0498-100L0498-100Liquid100 ml1 * 100 mLkr 93,00
- RPMI 1640 with stable glutamineRPMI 1640 with stable glutamineL0498-500Liquid500 ml1 * 500 mLkr 120,00
- RPMI 1640 with L-glutamine, with 25 mM HEPESRPMI 1640 with L-glutamine, with 25 mM HEPESL0495-500Liquid500 ml1 * 500 mLkr 150,00
- RPMI 1640 with stable glutamine, with 25 mM HEPESRPMI 1640 with stable glutamine, with 25 mM HEPESL0496-500Liquid500 ml1 * 500 mLkr 155,00
- RPMI 1640 without L-glutamine, with 25 mM HEPESRPMI 1640 without L-glutamine, with 25 mM HEPESL0490-500Liquid500 ml1 * 500 mLkr 150,00
- RPMI 1640 Dutch modification, without L-glutamine, with 1 g/l sodium bicarbonate, with 20 mM HEPESRPMI 1640 Dutch modification, without L-glutamine, with 1 g/l sodium bicarbonate, with 20 mM HEPESL0492-500Liquid500 ml1 * 500 mLkr 265,00
- RPMI 1640 without L-glutamine, without folic acidRPMI 1640 without L-glutamine, without folic acidL0503-500Liquid500 ml1 * 500 mL
- RPMI 1640 without L-glutamine, without phenol redRPMI 1640 without L-glutamine, without phenol redL0505-500Liquid500 ml1 * 500 mLkr 155,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, for 1 LRPMI 1640 with L-glutamine, without sodium bicarbonate, for 1 LP0860-N1LPowder11 * 1 Itemskr 380,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, for 5 LRPMI 1640 with L-glutamine, without sodium bicarbonate, for 5 LP0860-N5LP0860-N5LPowder11 * 1 Itemskr 850,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, for 10 lRPMI 1640 with L-glutamine, without sodium bicarbonate, for 10 lP0860-N10LPowder11 * 1 Itemskr 1 750,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, for 50 LRPMI 1640 with L-glutamine, without sodium bicarbonate, for 50 LP0860-N50LPowder11 * 1 Itemskr 1 975,00
- RPMI 1640 without L-glutamine, without sodium bicarbonate, for 1 lRPMI 1640 without L-glutamine, without sodium bicarbonate, for 1 lP0870-N1LP0870-N1LPowder11 * 1 Itemskr 345,00
- RPMI 1640 without L-glutamine, without sodium bicarbonate, for 10 LRPMI 1640 without L-glutamine, without sodium bicarbonate, for 10 LP0870-N10LP0870-N10LPowder11 * 1 Itemskr 1 700,00
- RPMI 1640 without L-glutamine, without sodium bicarbonate, for 50 LRPMI 1640 without L-glutamine, without sodium bicarbonate, for 50 LP0870-N50LP0870-N50LPowder11 * 1 Itemskr 2 025,00
- RPMI 1640 without L-glutamine, without sodium bicarbonate, without phenol red, for 1 lRPMI 1640 without L-glutamine, without sodium bicarbonate, without phenol red, for 1 lP0871-N1LP0871-N1LPowder11 * 1 Itemskr 345,00
- RPMI 1640 without L-glutamine, without sodium bicarbonate, without phenol red, for 10 LRPMI 1640 without L-glutamine, without sodium bicarbonate, without phenol red, for 10 LP0871-N10LP0871-N10LPowder11 * 1 Itemskr 1 700,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, with 25 mM HEPES, without phenol red, for 1 lRPMI 1640 with L-glutamine, without sodium bicarbonate, with 25 mM HEPES, without phenol red, for 1 lP0876-N1LPowder11 * 1 Itemskr 400,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, with 25 mM HEPES, without phenol red, for 10 lRPMI 1640 with L-glutamine, without sodium bicarbonate, with 25 mM HEPES, without phenol red, for 10 lP0876-N10LPowder11 * 1 Itemskr 1 850,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, with 25 mM HEPES, without phenol red, for 50 lRPMI 1640 with L-glutamine, without sodium bicarbonate, with 25 mM HEPES, without phenol red, for 50 lP0876-N50LP0876-N50LPowder11 * 1 Items
- RPMI 1640 with L-glutamine, without sodium bicarbonate, without phenol red, for 1 lRPMI 1640 with L-glutamine, without sodium bicarbonate, without phenol red, for 1 lP0880-N1LP0880-N1LPowder11 * 1 Itemskr 370,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, without phenol red, for 10 lRPMI 1640 with L-glutamine, without sodium bicarbonate, without phenol red, for 10 lP0880-N10LP0880-N10LPowder11 * 1 Itemskr 1 725,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, without phenol red, for 50 LRPMI 1640 with L-glutamine, without sodium bicarbonate, without phenol red, for 50 LP0880-N50LP0880-N50LPowder11 * 1 Itemskr 2 000,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, without glucose, for 1 lRPMI 1640 with L-glutamine, without sodium bicarbonate, without glucose, for 1 lP0883-N1LP0883-N1LPowder11 * 1 Itemskr 375,00
- RPMI 1640 with L-glutamine, without sodium bicarbonate, without glucose, for 10 LRPMI 1640 with L-glutamine, without sodium bicarbonate, without glucose, for 10 LP0883-N10LP0883-N10LPowder11 * 1 Itemskr 1 700,00