Active pirani gauges
Model APG.
These gauges have a measuring range from atmosphere to 1×10⁻⁴ mbar and combine the gauge head and controller in one compact active unit.
- Wide range, regulated, internal power supply - runs from standard DC power supplies 20 to 36 V
- Tolerant to voltage fluctuations
- Medium and low-pressure versions available - APG-M and -MP from atmosphere to 10⁻³ mbar. APG-L from atmosphere to 10⁻⁴ mbar
- Standard analogue output 0 to 10 V - Easy to interface with a computer or plc. Fault output indication
- Adjustable set-point for process control and interlocking
- Provides vacuum status indication
- Set-point level read visually or with voltmeter
- Catalog No:
- 619-0221
- 619-0220
- Model:
- APG-L-NW16
- APG-M-NW16
- Accuracy:
- 1×10ˉ4 mbar
- 1×10ˉ3 mbar
- Permitted ambient temperature for operation:
- +10...+60 °C
- +10...+60 °C
- Permitted ambient temperature for storage:
- 0...+70 °C
- 0...+70 °C
- Power input:
- 20 to 36 V dc (max 1 V ripple)
- 20 to 36 V dc (max 1 V ripple)
- Pk
- Active pirani gauges, APG-M-NW16: D02171000: EDWAD02171000
Specifications
ModelAPG-M-NW16Environmentally PreferableAccuracy1×10ˉ3 mbarPermitted ambient temperature for operation+10...+60 °CPermitted ambient temperature for storage0...+70 °CPower input20 to 36 V dc (max 1 V ripple) - Active pirani gauges, APG-L-NW16: D02173000: EDWAD02173000
Specifications
ModelAPG-L-NW16Environmentally PreferableAccuracy1×10ˉ4 mbarPermitted ambient temperature for operation+10...+60 °CPermitted ambient temperature for storage0...+70 °CPower input20 to 36 V dc (max 1 V ripple)