Sample loops for Cheminert® valves, PEEK
- BeskrivelseMaterialeKapasitetTrykkFp
- JOURJR-SLC-85-5JR-SLC-85-5Sample loopPEEK5 µlmax. 400 bar11 * 1 Itemskr 1 500,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-10JR-SLC-85-10Sample loopPEEK10 µlmax. 400 bar11 * 1 Itemskr 1 500,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-20JR-SLC-85-20Sample loopPEEK20 µlmax. 400 bar11 * 1 Itemskr 1 500,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-50JR-SLC-85-50Sample loopPEEK50 µlmax. 400 bar11 * 1 Itemskr 1 500,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-100JR-SLC-85-100Sample loopPEEK100 µlmax. 400 bar11 * 1 Itemskr 1 775,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-200JR-SLC-85-200Sample loopPEEK200 µlmax. 240 bar11 * 1 Itemskr 1 775,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-250JR-SLC-85-250Sample loopPEEK250 µlmax. 240 bar11 * 1 Itemskr 1 750,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-300JR-SLC-85-300Sample loopPEEK300 µlmax. 240 bar11 * 1 Itemskr 1 775,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-500JR-SLC-85-500Sample loopPEEK500 µlmax. 240 bar11 * 1 Itemskr 2 225,00
Specifications
Environmentally Preferable - JOURJR-SLC-85-1000JR-SLC-85-1000Sample loopPEEK1000 µlmax. 240 bar11 * 1 Itemskr 2 575,00
Specifications
Environmentally Preferable