ESD safety shoes, lace-up, uvex motion light, 6983
Leverandør: Uvex
uvex motion light
114-4888EA 1275 NOK
114-4888 114-4887 114-4890
ESD safety shoes, lace-up, uvex motion light, 6983
Sko og støvler
Wide, sports-fit ESD safety shoes made from nubuck leather upper, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. 120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Width: 11
Sertifiseringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRC