Micro pipette tips, solid phase extraction, HyperSep™
Leverandør: Thermo Scientific
HyperSep™
HYPE60109-211EA 3050 NOK
HYPE60109-211 HYPE60109-210 HYPE60109-212 HYPE60109-204 HYPE60109-205
Micro pipette tips, solid phase extraction, HyperSep™
HyperSep™ Tip is a revolutionary micro pipette tip for sample preparation in which the chromatographic media is directly attached to the inner surface of the tip without using polymers or glue. This avoids potential problems with contamination or permeability and maximises the surface area of the chromatographic media in contact with the sample.
  • Ideal when working with small samples
  • Tips contain just the desired chromatography material and nothing else
  • Available with a range of materials to suit a wide range of applications