Safety shoes, lace-up, Protektor Line, 1610
Leverandør: Abeba
ABEB1610/40EA 770 NOK
ABEB1610/40 ABEB1610/41 ABEB1610/42 ABEB1610/43 ABEB1610/44 ABEB1610/45 ABEB1610/46 ABEB1610/47 ABEB1610/36 ABEB1610/37 ABEB1610/48 ABEB1610/38 ABEB1610/39
Safety shoes, lace-up, Protektor Line, 1610
Anti-static shoes made from smooth leather upper and a PU sole.
  • Leather lining
  • PU sole with high slip resistance
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Suitable for use in kitchens and in food processing industries.
Colour: White
Sertifiseringer: EN ISO 20345:2011; S3, SRC