Du søkte etter: Waves and Oscillations Learning Activities
17  treff i ditt søk
SearchResultCount:"17"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Artikkenummer: (EDSC241-513)
Produsent: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: The Speed of Sound Kit is extremely simple to set up and use, and provides accurate and precise measurements for the speed of sound in air. The Edulab...
UOM: 1 * 1 Items
Artikkenummer: (765-0596)
Produsent: ELWE LEHRGERAETEBAU KLINGENTHA
Beskrivelse: Cone-shaped pressure chamber speaker with high power resistance. Used for general acoustic tests, eg together with resonator 765-0080. Connects to a t...
UOM: 1 * 1 Items
Artikkenummer: (765-1013)
Produsent: KVANT LTD.,MFF UK
Beskrivelse: Equipment for laser sound transmission experiments. The included microphone, tape recorder or function generator is used as the sound source. The rate...
UOM: 1 * 1 Items
Artikkenummer: (EDSC3032)
Produsent: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: The stroboscopic lamp is fitted with an extra-bright 3W LED, which is synchronised with the surface-wave generator. The control unit is equipped with ...
UOM: 1 * 1 Items
Artikkenummer: (BINHU56001)
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Vibration generator for exciting oscillating and waves mechanically, e.g. in coil springs, a rubber cord, a wire ring or a Chladni plate. In robust pl...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (BINHU10115)
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Suitable as a source of intense, high frequency sound, for example, for producing stationary sound waves in Kundt's tube.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Educational Materials, Waves and Oscillations, Applikasjon: Fysikk, Chladni plate, square, steel, 180×180 mm
Artikkenummer: (BINH1009957)
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Function generator with external sweep function and power amplifier for use in versatile student and practical experiments covering simple harmonic os...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (765-0154)
Produsent: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Educational Materials, Waves and Oscillations, Applikasjon: Fysikk, Monochord
UOM: 1 * 1 Each
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (BINHU8432770)
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Ideal sound source for acoustics experiments in the frequency range 60 Hz to 23 kHz.
Power capacity: 100 W (as per IEC 268-5)
Frequency ...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (BINHU55001)
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Large tuning fork for demonstration the vibrating legs of the fork.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (765-0157)
Produsent: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Metal frame with tension spring and membrane of Papyrolin. With the help of two tambourines and a pendulum ball, experiments with pressure waves can b...
UOM: 1 * 1 Each
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (765-1177)
Produsent: PHYWE
Beskrivelse: Quincke interference tube for determining wavelengths and frequencies by interfering with sound waves in air. Consists of two U-shaped metal tubes sli...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (765-0156)
Produsent: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: For acoustic detection of standing sound waves and for determining the wavelength of the sound source and then the speed of sound in air (Velocity=fre...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (765-0108)
Produsent: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Educational Materials, Waves and Oscillations, Applikasjon: Fysikk, Club for the tambourine
UOM: 1 * 1 Each
Market Source Item This is a MarketSource item. Additional charges may apply
Artikkenummer: (BINHU8431030)
Produsent: Binhold, Paul Lehrmittel
Beskrivelse: Tuning fork that allows for plotting oscillations on a sheet of paper. The oscillation is triggered by pushing the prongs of the fork together. The os...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 17
no targeter for Bottom