Safety shoes, lace-up, Envio
Leverandør: Honeywell Safety Products
DALL6543014/44EA 2550 NOK
DALL6543014/44
Safety shoes, lace-up, Envio
100% metal-free safety shoes made from derby, smooth full grain leather upper and a mono-density PU sole.
  • Fully lined with leather
  • Perforated leather insole, with shock-absorbent heel
  • Metal-free, puncture resistant midsole
  • Light Smartium-injected polymer toecap
Colour: Black
Sertifiseringer: EN ISO 20345:2011; S3, CI, SRC