Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Weather Stations
Rain gauge
Rain gauge
Rain gauge
612-0595
:
GLASWARENFABRIK KARL HECHT
Rain gauge
612-0595
:
GLASWARENFABRIK KARL HECHT
: 44278010
: HECH44278010
kr 185,00
1 * 1 Items
-
+
Description:
Rain gauge, 1 mm graduation, height 250 mm
Pk:
1
This rain gauge is made of clear plastic.
Easily readable scale
Holder for easier removing