First-aid kit, DIN 13169-E
Leverandør: Hartmann
HART7434099EA 0 NOK
HART7434099
First-aid kit, DIN 13169-E
Førstehjelpsutstyr Førstehjelpssett
This DIN 13169-E first aid kit is packed in a robust case made of sheet steel. It is suitable for 51 to 300 employees in administrative and commercial companies, for 21 to 100 employees in manufacturing and processing companies or for 11 to 50 employees on construction sites.
  • With rubber seal
  • Each case is individually packed in a cardboard box
  • Content is the same as for two DIN 13157 first aid kits
Contents:
2 × adhesive plasters DIN 13019-A, 5 m × 2,5 cm
16 × wound dressings DIN 13019-E, 10×6 cm
8 × fingertip plasters
8 × finger plasters, 12×2 cm
8 × plaster strips, 19×72 mm
16 × plaster strips, 25×72 mm
2 × wound dressing pack DIN 13151-K
6 × field dressing packs DIN 13151-M
2 × wound dressing pack DIN 13151-G
2 × Bandage DIN 13152-A, 60×80 cm
12 × compresses, 10×10 cm
4 × eye pads, 56×70 mm
2 × cooling compress
2 × emergency blanket, 210×160 cm
4 × bandages DIN 61634-FB, 6 cm
4 × bandages DIN 61634-FB, 8 cm
4 × triangular bandages DIN 13168-D
4 × foil bags, 30×40 cm
10 × non woven wipes, 20×30 cm
8 × medical disposable gloves DIN EN 455, large
1 × first aid booklet
1 × table of contents
1 pair of scissors DIN 58279-B, 19 cm