Floor signs, DuraStripe®, X-Treme, corner
: ERGOMAT
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe® program makes it the only total floor marking solution available on the market today. Economical than painting the floors.
- Ideal for smooth or newer floors but can be used on old floors as well
- Simple and easy peel-&-stick installation
- Easy to remove for a quick reconfiguration and hence a great Lean Manufacturing tool
- No dry time, cure time, fumes or messy clean up to limit productivity
- Available in 11 different colours
Made for critical environments and area where pivoting with forklifts / tow motors can occur. It has a special coated surface that facilitates easy cleaning.
Corner
- MaterialDimensionsPk
- Floor marking corner, X-Treme, yellowFloor marking corner, X-Treme, yellowERGODSXCR4YDSXCR4YPVC100×50 mm mm751 * 75 Itemskr 1 375,00: DSXCR4Y: ERGODSXCR4Y
Specifications
Environmentally Preferable - Floor marking corner, X-Treme, blueFloor marking corner, X-Treme, blueERGODSXCR4BDSXCR4BPVC100×50 mm601 * 60 Itemskr 1 375,00: DSXCR4B: ERGODSXCR4B
Specifications
Environmentally Preferable
Steps
- MaterialDimensionsPk
- Floor marking steps, X-Treme, blueFloor marking steps, X-Treme, blueERGO4070058XF9BL4070058XF9BLPVC240×90 mm251 * 25 Itemskr 1 200,00: 4070058XF9BL: ERGO4070058XF9BL
Specifications
Environmentally Preferable - Floor marking steps, X-Treme, yellowFloor marking steps, X-Treme, yellowERGODSXSTEPYDSXSTEPYPVC230×80 mm11 * 1 Itemskr 1 200,00: DSXSTEPY: ERGODSXSTEPY
Specifications
Environmentally Preferable - Floor marking steps, X-Treme, yellowFloor marking steps, X-Treme, yellowERGO4070058XF9Y4070058XF9YPVC240×90 mm251 * 25 Itemskr 1 200,00: 4070058XF9Y: ERGO4070058XF9Y
Specifications
Environmentally Preferable
T-Shapes
- MaterialDimensionsPk
- Floor marking T shape, X-Treme, blueFloor marking T shape, X-Treme, blueERGODSXTR4BDSXTR4BPVC100×150×50 mm501 * 50 Itemskr 1 375,00: DSXTR4B: ERGODSXTR4B
Specifications
Environmentally Preferable - Floor marking T shape, X-Treme, yellowFloor marking T shape, X-Treme, yellowERGODSXTR4YDSXTR4YPVC100×150×50 mm401 * 40 Itemskr 1 375,00: DSXTR4Y: ERGODSXTR4Y
Specifications
Environmentally Preferable