PP, non coated, non sterile, chromatography tested and certified. Manufactured from ultra-low bleed high purity basis resin, and fully lot tested by GC-MS for organic extractables. Designed for sensitive applications, lower sample concentrations, regulated laboratories, and/or TIC and full chromatograms.
- Microplates are chemically and thermally resistant and will tolerate temperatures from −80 to +121 °C
- SBS and ANSI standard footprint design for broad instrument compatibility
- Ideal for pharmaceutical applications, sample collection and storage, combinatorial chemistry and HTS applications
- U- and V-bottom well format for optimal sample recovery