Steady-Luc™ Firefly HTS assay kit
Leverandør: Biotium
30028-TEA 880 NOK
30028-T 99923. 30028-2 30028-3 30028-1
Steady-Luc™ Firefly HTS assay kit
Assay Cellulær analyse
Firefly luciferase is widely used as a transcriptional reporter gene for research and drug screening. The Steady-Luc™ HTS kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening.