Culture media, ready to use, in plates
Supplier: THERMO FISHER DIAGNOSTICS
New Product
Certificates
About this item
Product Information
- Catalog #Supplier #StandardsPkAvailabilityPrice
- MacConkey No 3 agar
- TSA tryptic soy agar with sheep bloodTSA tryptic soy agar with sheep bloodOXOIPB5012APB5012A-10Log in for availability1 * 10 Itemskr 210,00
- Neisseria selective medium
- Sorbitol MacConkey agar with CefiximeSorbitol MacConkey agar with CefiximeOXOIPO0702APO0702A-10Log in for availability1 * 10 Itemskr 315,00
- Sabouraud glucose agar
- Columbia Aescul./Macconkey Mod. agarColumbia Aescul./Macconkey Mod. agarOXOIPB5224EPB5224E-10Log in for availability1 * 10 Itemskr 255,00
- Tryptone Soya agar with Lecithine, triple wrappedTryptone Soya agar with Lecithine, triple wrappedOXOIPO5501BPO5501BBP, EP, JP, USP20Log in for availability1 * 20 Itemskr 3 650,00
- Blood agar/ Chocolate Duoplate
- MacConkey No.3/Blood agar
- Schaedler/Schaedler KV
- Enterohaemolysin agar with bloodEnterohaemolysin agar with bloodOXOIPB5105APB5105A-10Log in for availability1 * 10 Itemskr 360,00
- SS Salmonella Shigella agar
- Listeria Oxford agar
- Nutrient agar
- TSA tryptic soy agar
- Gassner agar
- Schaedler-K/V agar with blood
- Legionella BCYE medium with antibioticsLegionella BCYE medium with antibioticsOXOIPO5325APO5325A-10Log in for availability1 * 10 Itemskr 480,00
- CLED agar
- Desoxycholate agar
- Mueller-Hinton agar
- TSA tryptic soy agar
- TSA tryptic soy agar, irradiatedTSA tryptic soy agar, irradiatedOXOIPO5012BPO5012B-10Log in for availability1 * 10 Itemskr 360,00
- TSA tryptic soy agar
- Slanetz & Bartley Enterococci agarSlanetz & Bartley Enterococci agarOXOIPO5018APO5018ADIN EN ISO 7899-210Log in for availability1 * 10 Itemskr 185,00
- MacConkey without NaCl
- Anaerobic blood agar (Wilkins Chalgren) with nalidixic acid and Tween™, 10×90 mmAnaerobic blood agar (Wilkins Chalgren) with nalidixic acid and Tween™, 10×90 mmOXOIPB0113APB0113A-10Log in for availability1 * 10 Itemskr 260,00
- Brilliance ESBL agar
- MRS-TKTC agar
- Burkholderia cepacia agar
- Rose Bengal Chlor agar
- Chromogenic UTI Clarity agar
- Yersinia agar (CIN)
- Karmali selective agar
- Mueller Hinton agar
- R2A agar
- Columbia agar 5% sheep blood
- MYP agar
- Pseudomonas CFC agar
- HTM agar
- Braziers Clostridium Diff Sel agarBraziers Clostridium Diff Sel agarOXOIPB1055APB1055A-10Log in for availability1 * 10 Itemskr 335,00
- TSA with Lecithin, triple wrappedTSA with Lecithin, triple wrappedOXOIPO5511DPO5511D-100Log in for availability1 * 100 Itemskr 4 400,00
- GSP selective agar
- Saboraud Dextr/Chromog.C. albicansSaboraud Dextr/Chromog.C. albicansOXOIPO5243EPO5243E-20Log in for availability1 * 20 Itemskr 305,00
- Wort agar
- Karmali/Karmali medium
- Tergitol-7-Lactose TTC agar 55 mmTergitol-7-Lactose TTC agar 55 mmOXOIPO5411JPO5411J-10Log in for availability1 * 10 Itemskr 205,00
- DST agar
- Dematophyte selective agar acc. TaplinDematophyte selective agar acc. TaplinOXOIPO5087APO5087A-10Log in for availability1 * 10 Itemskr 305,00
- Rice agar
- XLD/Salmonella Chromogenic DuoplateXLD/Salmonella Chromogenic DuoplateOXOIPO5248EPO5248E-10Log in for availability1 * 10 Itemskr 335,00
- E.Coli/Coliform Chromogen agar
- CLED/ MacConkey No.3 agar
- XLD agar
- Bacillus cereus agar (PEMBA)Bacillus cereus agar (PEMBA)OXOIPO5048APO5048ADIN 10198-110Log in for availability1 * 10 Itemskr 270,00
- Iso-Sensitest agar, 10×90 mm
- Malt extract agar
- Kanamycin aesculin azide agar
- Chromogenic Candida agar
- Chromogenic C. albicans agar
- MRS agar
- TSA agar with inhibitor
- TSYE agar
- Yeast glucose Chloramphenicol agarYeast glucose Chloramphenicol agarOXOIPO5032APO5032A-10Log in for availability1 * 10 Itemskr 215,00
- Sabouraud dextrose agar
- M-CP agar
- Brilliance MRSA 2 agar, 10×90 mmBrilliance MRSA 2 agar, 10×90 mmOXOIPO5310APO5310A-10Log in for availability1 * 10 Itemskr 405,00
- SS agar/XLD agar Duoplate
- Mycoplasma / Ureaplasma agar
- Legionella GVPC agar
- TSA tryptic soy agar
- Sabouraud dextrose agar
- PCA Plate count agar
- VRB (Violet red bile) agarVRB (Violet red bile) agarOXOIPO5075APO5075AISO 483210Log in for availability1 * 10 Itemskr 185,00
- Violet red bile glucose agarSpecifications:New ProductNew Product
- Sabouraud glucose Chloramphenicol agarSabouraud glucose Chloramphenicol agarOXOIPO5070APO5070A-10Log in for availability1 * 10 Itemskr 250,00
- Legionella BCYE agar
- Legionella MWY selective agar
- Enterococci Bile Aesculin Azide agarEnterococci Bile Aesculin Azide agarOXOIPO5062APO5062A-10Log in for availability1 * 10 Itemskr 190,00
- Chocolate agar with Vitox
- Legionella BCYE without L-CysteineLegionella BCYE without L-CysteineOXOIPO5028APO5028A-10Log in for availability1 * 10 Itemskr 485,00
- Mannitol salt agar
- Pseudomonas agar (Cetrimide)Pseudomonas agar (Cetrimide)OXOIPO5076APO5076ADIN 1278010Log in for availability1 * 10 Itemskr 235,00
- Biggy agar
- Mueller Hinton with blood
- Campylobacter agar (Skirrow)
- Hektoen enteric agar
- Salmonella Chromogenic medium IISalmonella Chromogenic medium IIOXOIPO5098APO5098A-10Log in for availability1 * 10 Itemskr 385,00
- Dermasel selective agar
- Campylobacter agar (Butzler)
- Schaedler agar with blood
- Aesculin blood agar mod.
- VRB (Violet red bile) agar with MUGVRB (Violet red bile) agar with MUGOXOIPO5031APO5031A-10Log in for availability1 * 10 Itemskr 270,00
- Dichloran Bengalrot Chloramphenicaol agarDichloran Bengalrot Chloramphenicaol agarOXOIPO1227APO1227A-10Log in for availability1 * 10 Itemskr 250,00
- MLCB agar
- Legionella BMPA selective agar
- Haemophilus selective agar
- Brilliance Staph 24 agar, 10×90 mmBrilliance Staph 24 agar, 10×90 mmOXOIPO1186APO1186A-10Log in for availability1 * 10 Itemskr 540,00
- Sabouraud glucose Chloramphenicol gentamicin agarSabouraud glucose Chloramphenicol gentamicin agarOXOIPO5096APO5096A-10Log in for availability1 * 10 Itemskr 245,00
- Eosin methylene blue agar (mod.) LevineEosin methylene blue agar (mod.) LevineOXOIPO5045APO5045A-10Log in for availability1 * 10 Itemskr 180,00
- TSA agar with neutralisatorTSA agar with neutralisatorOXOIPO5095APO5095ABP/EP/JP/USP10Log in for availability1 * 10 Itemskr 170,00
- Helicobacter Pylori selective agar, 19 mlHelicobacter Pylori selective agar, 19 mlOXOIPB0398APB0398A-10Log in for availability1 * 10 Itemskr 540,00
- Iso-Sensitest agar with blood +2% NaclIso-Sensitest agar with blood +2% NaclOXOIPB5003APB5003A-10Log in for availability1 * 10 Itemskr 215,00
- VRE selective agar
- Campylobacter blood-free CCDA agarCampylobacter blood-free CCDA agarOXOIPO5091APO5091AISO 1027210Log in for availability1 * 10 Itemskr 255,00
- Dermatophyte medium with Phenol RedDermatophyte medium with Phenol RedOXOIPO0166APO0166A-10Log in for availability1 * 10 Itemskr 435,00
- Blood agar No.2 with sheep bloodBlood agar No.2 with sheep bloodOXOIPB0115APB0115A-10Log in for availability1 * 10 Itemskr 200,00
- Baird Parker agar
- Potato extract glucose agarPotato extract glucose agarOXOIPO0186APO0186ABP/EP/JP/USP10Log in for availability1 * 10 Itemskr 270,00
- Pasteurella Selective agar
- Columbia sheep blood agar
- Edwards agar modified
- Brilliance™ CampyCount agar
- TBX medium
- Staph Strep agar
- Listeria Palcam agarListeria Palcam agarOXOIPO5104APO5104ACEN ISO/TS 11133-210Log in for availability1 * 10 Itemskr 275,00
- Gardnerella Vaginalis Select. agar 2X10Gardnerella Vaginalis Select. agar 2X10OXOIPB5067APB5067A-10Log in for availability1 * 10 Itemskr 310,00
- Bordetella agar