Photometers monoparameter
Leverandør: Palintest
The Chlorometer Duo range of photometers are portable handheld instruments ideal for dual range chlorine analysis for routine water monitoring and other applications. It is used to test the potability of drinking water either by DPA method or by measuring higher levels of chlorine concentration in the water samples.
  • Simple operation - blank or measure using a single button
  • Ideal for field use - waterproof and battery powered
  • Available in both hard (PTH) and soft (PTS) carry case kits
BESTILL
Utvid alle / Skjul alle
Beskrivelse Område Chemical method Type Fp Art.nr Antall
Photometer, hard carry case kit 0,01 - 5 mg/l (DPD)
1 - 250 mg/l HR tablets
DPD Chlorometer Duo 1 PALIPTH027
Photometer, soft carry case kit 0,01 - 5 mg/l (DPD)
1 - 250 mg/l HR tablets
DPD Chlorometer Duo 1 PALIPTS027
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
romtemp ved drift (°C) 0...50
Automatisk switch-off
Display LCD (128×64 pixel screen)
Modell Chlorometer Duo
Optikk dual LED light source optical system with narrow band (10 nm) wavelength filters and photodetectors
Strømforsyning 2 AA batteries 1,5 V
Prøvekammer Waterproof
Vekt (kg) 0, 2 (including batteries)
B×D×H (mm) 150×65×40
TILBEHØR
Utvid alle / Skjul alle
Accessories
Beskrivelse Fp Art.nr Antall
NDF validation standards for Chlorometer Duo 1 PALIPTC027
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall