Acrylamide/ Bisacrylamide
Leverandør: G-Biosciences
Acrylamide/bisacrylamide solutions are available at the most common ratios (37.5:1 or 29:1) for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bisacrylamide. Supplied as 40% solutions prepared from electrophoresis grade acrylamide and bis-acrylamide in ultra-pure water.
BESTILL
Utvid alle / Skjul alle
Beskrivelse Fp Art.nr Antall
Acrylamide, 40% solution 500 ml 786-508
Bisacrylamide, 2% solution 500 ml 786-509
Acrylamide/bisacrylamide (37.5:1), 40% solution 500 ml 786-502
- 40 g 786-503
Acrylamide/bisacrylamide (37.5:1), premixed powder 200 g 786-504
- 500 ml 786-505
Acrylamide/bisacrylamide (29:1), premixed powder 40 g 786-506
Acrylamide/bisacrylamide (29:1), premixed powder 200 g 786-507
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall