Glove liners, HyFlex® 48-100 / 48-101
Supplier: Ansell
Certificates
About this item
These knitted nylon gloves with PU coated palm are designed for light applications that require good levels of comfort and abrasion resistance.
- Optimum protection for light tasks, high level of comfort
- Good freedom of movement for precise handling of small objects
- Elasticated nylon liner for exceptionally close fit
- Excellent alternative to light leather gloves
- Available in white or black
Ideal for the precise handling of small objects in dry or light oily environments.
Colour: White
EN 388 (003X), Cat. II
Certifications: EN 388:2016 (4131A); Cat. II
Specifications
- Catalog No:
- ANSE48-100/7
- ANSE48-100/8
- ANSE48-101/8
- ANSE48-100/10
- ANSE48-101/9
- ANSE48-100/11
- ANSE48-100/6
- ANSE48-101/10
- ANSE48-101/11
- ANSE48-101/6
- ANSE48-101/7
- ANSE48-100/9
- Model:
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
Specifications
Product Information
- Catalog #Supplier #ModelGlove sizeColourLengthPackedPkAvailabilityPrice
- ANSE48-100/648-100/648-1006White210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 530,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-100/748-100/748-1007White210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 530,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-100/848-100/848-1008White210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 530,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-100/948-100/948-1009White210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 530,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-100/1048-100/1048-10010White210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 530,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-100/1148-100/1148-10011White210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 530,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-101/648-101/648-1006Black210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 500,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-101/748-101/748-1007Black210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 500,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-101/848-101/848-1008Black210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 500,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-101/948-101/948-1009Black210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 500,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-101/1048-101/1048-10010Black210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 500,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A) - ANSE48-101/1148-101/1148-10011Black210 - 265 mm12 pairs/bag12 PAIRLog in for availability1 * 12 PARkr 500,00Specifications:
More
Specifications
Model48-100Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialNylonGauge13Cuff styleKnit wristEN normEN 388:2016 (4131A)