Norway
Order Entry
Forma™ Racks for Ultra Low Temperature Freezers
Forma™ Racks for Ultra Low Temperature Freezers
Supplier:  Thermo Scientific
Certificates
undefined
Forma™ Racks for Ultra Low Temperature Freezers
Supplier:  Thermo Scientific
Product Information
  • Catalog #
    Supplier #
    Compartments
    Colour code
    For
    Cryobox height
    W×D×H ext.
    Availability
    Price
  • Side access rack, adjustable, for 490 and 651 L capacity upright freezers
    Side access rack, adjustable, for 490 and 651 L capacity upright freezers
    HERA920018
    920018
    20
    Silver
    17,3 and 23 cu. ft. capacity upright freezers
    50 mm
    140×559×277 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 025,00
    kr 3 025,00
    1 * 1 Items
       kr 3 025,00
    Supplier Article Number: 920018
    Local Article Number: HERA920018
    Specifications:
    Specifications
    Environmentally Preferable
  • Side access rack, adjustable, for 490 and 651 L capacity upright freezers, stainless steel, 5 shelves
    Side access rack, adjustable, for 490 and 651 L capacity upright freezers, stainless steel, 5 shelves
    HERA920022
    920022
    22
    Silver
    17,3 and 23 cu. ft. capacity upright freezers
    50 mm
    135×625×295 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 025,00
    kr 3 025,00
    1 * 1 Items
       kr 3 025,00
    Supplier Article Number: 920022
    Local Article Number: HERA920022
    Specifications:
    Specifications
    Environmentally Preferable
  • Side access rack, adjustable, for 490 and 651 L capacity upright freezers, stainless steel, 3 shelves
    Side access rack, adjustable, for 490 and 651 L capacity upright freezers, stainless steel, 3 shelves
    HERA920017
    920017
    12
    Silver
    17,3 and 23 cu. ft. capacity upright freezers
    76,2 mm
    137×559×295 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 025,00
    kr 3 025,00
    1 * 1 Items
       kr 3 025,00
    Supplier Article Number: 920017
    Local Article Number: HERA920017
    Specifications:
    Specifications
    Environmentally Preferable
  • Sliding drawer rack, for 490 and 651 L capacity upright freezers, 3 shelves
    Sliding drawer rack, for 490 and 651 L capacity upright freezers, 3 shelves
    HERA920096
    920096
    20
    Silver
    17,3 and 23 cu. ft. capacity upright freezers
    50 mm
    140×302×559 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 920096
    Local Article Number: HERA920096
    Specifications:
    Specifications
    Environmentally Preferable
  • Sliding drawer rack, for 490 and 651 L capacity upright freezers, stainless steel, 3 shelves
    Sliding drawer rack, for 490 and 651 L capacity upright freezers, stainless steel, 3 shelves
    HERA920097
    920097
    12
    Silver
    17,3 and 23 cu. ft. capacity upright freezers
    76,2 mm
    140×559×302 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 920097
    Local Article Number: HERA920097
    Specifications:
    Specifications
    Environmentally Preferable