Norway
Order Entry
VWR®, Cell Strainers
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
undefined
VWR®, Cell Strainers
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 732-2757
  • 732-2759
  • 732-2758
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Cell strainers, 40 µm pore size, blue frame
    Cell strainers, 40 µm pore size, blue frame
    732-2757
    50
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 1 650,00
    kr 1 650,00
    1 * 50 Items
       kr 1 650,00
    Local Article Number: VWRI732-2757
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Cell strainers, 100 µm pore size, yellow frame
    Cell strainers, 100 µm pore size, yellow frame
    732-2759
    50
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 1 650,00
    kr 1 650,00
    1 * 50 Items
       kr 1 650,00
    Local Article Number: VWRI732-2759
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Cell strainers, 70 µm pore size, white frame
    Cell strainers, 70 µm pore size, white frame
    732-2758
    50
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 1 650,00
    kr 1 650,00
    1 * 50 Items
       kr 1 650,00
    Local Article Number: VWRI732-2758
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact