Spectrophotometric colorimeters, model AF, Lovibond®
Catalog # TINT107103
Leverandør: Lovibond Tintometer
Spesifikasjoner
- Lyskilde:2×12 Volt, 20 W tungsten halogen lamp
- Beskrivelse:AF710-3
- Modell:Model AF
- Lystyper:Approximates to daylight
- Optisk system:Visual colour comparison against colour glasses
- Vekt:8,3 kg
- B×D×H:330×100×480 mm
- Fp:1
- For:Oils and fats
Spesifikasjoner
Om dette produktet
The AF710-3 is a visual instrument used for colour grading of oils and fats.
Colour is determined by comparing light transmitted through a column of oil with that transmitted through a series of Red and Yellow glasses calibrated in accordance with the AOCS-Tintometer® Colour Scale.
The glasses enable a colour match to be found from the light passing through the oil. This colour is then expressed in units of Red and Yellow.