Anti-POLDIP3 Rabbit Polyclonal Antibody
Catalog # AVIVARP41048_P050
Supplier: Aviva Systems Biology
- Antibody type:Primary
- Antigen name:Polymerase (DNA-directed), delta interacting protein 3
- Antigen symbol:POLDIP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:84271
- Antigen synonyms:PDIP46|SKAR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: POLDIP3 (polymerase (DNA-directed), delta interacting protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: