Anti-A1CF Rabbit Polyclonal Antibody
AVIVARP41109_P050
- Antibody type:Primary
- Antigen name:APOBEC1 complementation factor
- Antigen symbol:A1CF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:29974
- Antigen synonyms:ACF|ACF64|ACF65|ASP|APOBEC1CF
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: A1CF (APOBEC1 complementation factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human