Anti-SLC25A35 Rabbit Polyclonal Antibody
AVIVARP44177_P050
- Antibody type:Primary
- Antigen name:solute carrier family 25, member 35
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:399512
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SLC25A35 (solute carrier family 25, member 35)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: