Anti-HNF4G Rabbit Polyclonal Antibody
AVIVARP45615_P050
- Antibody type:Primary
- Antigen name:hepatocyte nuclear factor 4, gamma
- Antigen symbol:HNF4G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3174
- Antigen synonyms:NR2A2|NR2A3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HNF4G (hepatocyte nuclear factor 4, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: