Anti-TMEM104 Rabbit Polyclonal Antibody
AVIVARP47331_T100
- Antibody type:Primary
- Antigen name:transmembrane protein 104
- Antigen symbol:TMEM104
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:54868
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: TMEM104 (transmembrane protein 104)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: