Norway
Order Entry
Anti-CDY1 Rabbit Polyclonal Antibody
  AVIVARP48645_T100
Anti-CDY1 Rabbit Polyclonal Antibody
  AVIVARP48645_T100
 : ARP48645_T100
 : AVIVARP48645_T100
 
kr 3 850,00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Chromodomain protein, Y-linked, 1
  • Antigen symbol:
    CDY1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    9085
  • Antigen synonyms:
    CDY|CDY1A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: CDY1 (chromodomain protein, Y-linked, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human