Anti-EFCAB13 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
AVIVARP52912_P050EA 4275 NOK
AVIVARP52912_P050
Anti-EFCAB13 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: EFCAB13 (EF-hand calcium binding domain 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR