Anti-PEPD Rabbit Polyclonal Antibody
AVIVARP56088_P050
- Antibody type:Primary
- Antigen name:peptidase D
- Antigen symbol:PEPD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:5184
- Antigen synonyms:PROLIDASE
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PEPD (peptidase D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: