Anti-PAFAH1B3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
AVIVARP56374_P050EA 4275 NOK
AVIVARP56374_P050
Anti-PAFAH1B3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PAFAH1B3 (platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR