Anti-E2F7 Goat Antibody

Leverandør: OriGene
ORIGTA305912EA 6325 NOK
ORIGTA305912
Anti-E2F7 Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: E2F7 (E2F transcription factor 7)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR