Anti-AGAP2 Goat Antibody

Leverandør: OriGene
ORIGTA305917EA 4400 NOK InStock
ORIGTA305917
Anti-AGAP2 Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: AGAP2 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more