Anti-CCR3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305938EA 4825 NOK InStock
ORIGTA305938
Anti-CCR3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-2?g/ml

Type: Primary
Antigen: CCR3 (chemokine (C-C motif) receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more