Anti-CRADD Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305944EA 8425 NOK
ORIGTA305944
Anti-CRADD Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CRADD (CASP2 and RIPK1 domain containing adaptor with death domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR