Anti-TNFRSF25 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305965EA 4825 NOK InStock
ORIGTA305965
Anti-TNFRSF25 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: TNFRSF25 (tumor necrosis factor receptor superfamily, member 25)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more