Anti-APAF1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305972EA 4825 NOK InStock
ORIGTA305972
Anti-APAF1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-2?g/ml; Immunochemistry: 1?g/ml

Type: Primary
Antigen: APAF1 (apoptotic peptidase activating factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more