Anti-DNASE2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305979EA 4825 NOK InStock
ORIGTA305979
Anti-DNASE2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: DNASE2 (deoxyribonuclease II, lysosomal)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more