Anti-CX3CL1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305991EA 8425 NOK
ORIGTA305991
Anti-CX3CL1 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 0.5-1 mg/ml

Type: Primary
Antigen: CX3CL1 (chemokine (C-X3-C motif) ligand 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR