Anti-IKBKB Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA305996EA 8425 NOK
ORIGTA305996
Anti-IKBKB Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR