Anti-APP Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306003EA 8425 NOK
ORIGTA306003
Anti-APP Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR