Anti-CXCR6 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306016EA 8425 NOK
ORIGTA306016
Anti-CXCR6 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR