Anti-ACIN1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306018EA 8425 NOK
ORIGTA306018
Anti-ACIN1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ACIN1 (apoptotic chromatin condensation inducer 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR