Anti-NOL3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306023EA 8425 NOK
ORIGTA306023
Anti-NOL3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: NOL3 (nucleolar protein 3 (apoptosis repressor with CARD domain))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR