Anti-TNFRSF10C Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306046EA 4825 NOK InStock
ORIGTA306046
Anti-TNFRSF10C Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more