Anti-CASP12 Rabbit Polyclonal Antibody
Leverandør: OriGene
ORIGTA306053EA 4825 NOK InStock
ORIGTA306053
Anti-CASP12 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CASP12 (caspase 12 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat