Anti-TLR4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306126EA 4825 NOK InStock
ORIGTA306126
Anti-TLR4 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 2.5-5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more